Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRSF4
Entrez Gene ID
614310
Gene Name
serine and arginine rich splicing factor 4
Gene Aliases
SFRS4
Chromosome Location
Chr.2: 124375055 - 124400705 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101231.1 NP_001094701.1 1 - 2 236 144
GenBank mRNA BC151318.1 - 1 - 2 236 144

View Full Product Details