Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C8H9orf85
Entrez Gene ID
614778
Gene Name
chromosome 8 C9orf85 homolog
Gene Aliases
-
Chromosome Location
Chr.8: 48157772 - 48227309 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101235.2 NP_001094705.1 1 - 1 149 101
GenBank mRNA BC148025.1 - 1 - 1 150 101

View Full Product Details