Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD19
Entrez Gene ID
615438
Gene Name
BTB domain containing 19
Gene Aliases
-
Chromosome Location
Chr.3: 101200430 - 101207033 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101241.1 NP_001094711.1 3 - 4 636 84
GenBank mRNA BC149224.1 - 3 - 4 636 84

View Full Product Details