Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFCAB2
Entrez Gene ID
615715
Gene Name
EF-hand calcium binding domain 2
Gene Aliases
-
Chromosome Location
Chr.16: 32201887 - 32315747 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101247.1 NP_001094717.1 6 - 7 409 103
GenBank mRNA BC149943.1 - 6 - 7 409 103

View Full Product Details