Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C26H10orf82
Entrez Gene ID
616318
Gene Name
chromosome 26 C10orf82 homolog
Gene Aliases
-
Chromosome Location
Chr.26: 37058422 - 37066152 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101253.2 NP_001094723.1 4 - 5 489 66
GenBank mRNA BC149249.1 - 4 - 5 490 66

View Full Product Details