Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXO4
Entrez Gene ID
618793
Gene Name
forkhead box O4
Gene Aliases
-
Chromosome Location
Chr.X: 79528188 - 79535459 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101277.1 NP_001094747.1 2 - 3 2014 118
GenBank mRNA BC148034.1 - 2 - 3 2014 118

View Full Product Details