Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSGALNACT1
Entrez Gene ID
528057
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
CHGN
Chromosome Location
Chr.27: 37895018 - 38233887 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102116.2 NP_001095586.1 7 - 8 1704 68
GenBank mRNA BC151604.1 - 7 - 8 1705 68

View Full Product Details