Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPM1N
Entrez Gene ID
530233
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1N (putative)
Gene Aliases
-
Chromosome Location
Chr.18: 53096288 - 53101654 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102128.1 NP_001095598.1 1 - 2 117 86
GenBank mRNA BC149371.1 - 1 - 2 117 86

View Full Product Details