Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD18
Entrez Gene ID
530484
Gene Name
abhydrolase domain containing 18
Gene Aliases
C17H4orf29
Chromosome Location
Chr.17: 29694439 - 29733173 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102129.2 NP_001095599.1 2 - 3 166 147
GenBank mRNA BC150053.1 - 2 - 3 167 147

View Full Product Details