Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFAND3
Entrez Gene ID
532641
Gene Name
zinc finger AN1-type containing 3
Gene Aliases
-
Chromosome Location
Chr.23: 11729171 - 12057183 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102145.1 NP_001095615.1 3 - 4 476 79
GenBank mRNA BC148099.1 - 3 - 4 476 79

View Full Product Details