Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCN5
Entrez Gene ID
534658
Gene Name
cellular communication network factor 5
Gene Aliases
WISP2
Chromosome Location
Chr.13: 73110493 - 73125825 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102176.1 NP_001095646.1 2 - 3 295 109
GenBank mRNA BT030745.1 - 2 - 3 295 109

View Full Product Details