Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCML4
Entrez Gene ID
535263
Gene Name
Scm polycomb group protein like 4
Gene Aliases
-
Chromosome Location
Chr.9: 42257455 - 42361821 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102184.2 NP_001095654.1 3 - 4 630 74
GenBank mRNA BC149641.1 - 3 - 4 633 74

View Full Product Details