Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGCB
Entrez Gene ID
535372
Gene Name
sarcoglycan beta
Gene Aliases
-
Chromosome Location
Chr.6: 67878832 - 67905909 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102188.1 NP_001095658.1 4 - 5 633 95
GenBank mRNA BC149178.1 - 4 - 5 633 95

View Full Product Details