Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EGLN2
Entrez Gene ID
536763
Gene Name
egl-9 family hypoxia inducible factor 2
Gene Aliases
-
Chromosome Location
Chr.18: 50047626 - 50060820 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102193.1 NP_001095663.1 2 - 3 1116 96
GenBank mRNA BT030693.1 - 2 - 3 1116 96

View Full Product Details