Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRAPPC11
Entrez Gene ID
537211
Gene Name
trafficking protein particle complex 11
Gene Aliases
C27H4orf41
Chromosome Location
Chr.27: 14375240 - 14414603 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102197.1 NP_001095667.1 24 - 25 2840 75
GenBank mRNA BC147917.1 - 24 - 25 2840 75

View Full Product Details