Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTD
Entrez Gene ID
537669
Gene Name
biotinidase
Gene Aliases
Biotinase
Chromosome Location
Chr.1: 152638136 - 152680280 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102206.1 NP_001095676.1 2 - 3 641 65
GenBank mRNA BC149584.1 - 2 - 3 641 65

View Full Product Details