Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDYL
Entrez Gene ID
539013
Gene Name
chromodomain Y like
Gene Aliases
-
Chromosome Location
Chr.23: 49586792 - 49745098 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102223.2 NP_001095693.1 4 - 5 1373 82
GenBank mRNA BC151603.1 - 4 - 5 1375 82
EF687906.1 - 6 - 7 1456 82
EF690280.1 - 4 - 5 1426 82
EF690281.1 - 3 - 4 759 82
EF690282.1 - 1 - 2 317 82

View Full Product Details