Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LRIF1
- Entrez Gene ID
-
539067
- Gene Name
- ligand dependent nuclear receptor interacting factor 1
- Gene Aliases
-
C3H1orf103
- Chromosome Location
-
Chr.3: 32407803 - 32428006 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC149364.1 |
- |
2 - 3 |
532 |
138 |
View Full Product Details