Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH18
Entrez Gene ID
539464
Gene Name
protocadherin 18
Gene Aliases
-
Chromosome Location
Chr.17: 20287884 - 20302108 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102229.2 NP_001095699.1 3 - 4 3114 93
GenBank mRNA BC151333.1 - 3 - 4 3115 93

View Full Product Details