Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PYGO2
Entrez Gene ID
540401
Gene Name
pygopus family PHD finger 2
Gene Aliases
-
Chromosome Location
Chr.3: 15627426 - 15631625 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102242.2 NP_001095712.1 1 - 2 277 62
GenBank mRNA BC151715.1 - 1 - 2 278 62

View Full Product Details