Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC12
Entrez Gene ID
615707
Gene Name
trafficking protein particle complex 12
Gene Aliases
TTC15
Chromosome Location
Chr.8: 111052948 - 111089546 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102290.1 NP_001095760.1 9 - 10 2001 96
GenBank mRNA BC147918.1 - 9 - 10 2001 96

View Full Product Details