Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF28
Entrez Gene ID
616969
Gene Name
Rho guanine nucleotide exchange factor 28
Gene Aliases
RGNEF
Chromosome Location
Chr.20: 7682926 - 8027701 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102307.1 NP_001095777.2 21 - 22 2920 61
GenBank mRNA BC149175.1 - 21 - 22 2920 61

View Full Product Details