Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCUR1
Entrez Gene ID
617129
Gene Name
mitochondrial calcium uniporter regulator 1
Gene Aliases
CCDC90A
Chromosome Location
Chr.23: 42868590 - 42890961 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102309.2 NP_001095779.1 4 - 5 796 142
GenBank mRNA BC149934.1 - 4 - 5 797 142

View Full Product Details