Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SURF4
Entrez Gene ID
526045
Gene Name
surfeit 4
Gene Aliases
-
Chromosome Location
Chr.11: 104267122 - 104278750 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102509.1 NP_001095979.1 3 - 4 481 88
GenBank mRNA BC151306.1 - 3 - 4 481 88

View Full Product Details