Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNMA8A
Entrez Gene ID
532062
Gene Name
PNMA family member 8A
Gene Aliases
PNMAL1
Chromosome Location
Chr.18: 53647296 - 53651183 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102512.1 NP_001095982.1 1 - 2 233 86
GenBank mRNA BC151672.1 - 1 - 2 233 86

View Full Product Details