Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM52B
Entrez Gene ID
618567
Gene Name
transmembrane protein 52B
Gene Aliases
C5H12orf59
Chromosome Location
Chr.5: 99782215 - 99792178 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102525.2 NP_001095995.1 1 - 2 508 60
GenBank mRNA BC151527.1 - 1 - 2 509 60

View Full Product Details