Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HHEX
Entrez Gene ID
539542
Gene Name
hematopoietically expressed homeobox
Gene Aliases
-
Chromosome Location
Chr.26: 14075882 - 14081683 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105424.1 NP_001098894.1 1 - 2 389 106
GenBank mRNA BC134632.1 - 1 - 2 389 106

View Full Product Details