Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNF5
Entrez Gene ID
616187
Gene Name
ring finger protein 5
Gene Aliases
-
Chromosome Location
Chr.23: 27209390 - 27211704 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105445.2 NP_001098915.1 4 - 5 348 121
GenBank mRNA BC114818.1 - 4 - 5 350 121

View Full Product Details