Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR6
Entrez Gene ID
526884
Gene Name
WD repeat domain 6
Gene Aliases
-
Chromosome Location
Chr.22: 50975111 - 50983679 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105623.1 NP_001099093.1 5 - 6 2918 91
GenBank mRNA BC153250.1 - 5 - 6 2918 91

View Full Product Details