Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA5L1
Entrez Gene ID
533070
Gene Name
spermatogenesis associated 5 like 1
Gene Aliases
-
Chromosome Location
Chr.10: 65036629 - 65057108 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105625.1 NP_001099095.1 3 - 4 1463 66
GenBank mRNA BC114696.1 - 3 - 4 1463 66

View Full Product Details