Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EWSR1
Entrez Gene ID
534073
Gene Name
EWS RNA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 68553309 - 68579071 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109800.2 NP_001103270.1 3 - 4 157 71
GenBank mRNA BC149284.1 - 3 - 4 121 71
BC153844.1 - 3 - 4 158 71
HQ011404.1 - 3 - 4 138 71

View Full Product Details