Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRAS
Entrez Gene ID
541140
Gene Name
KRAS proto-oncogene, GTPase
Gene Aliases
KRAS2, p21ras
Chromosome Location
Chr.5: 84749322 - 84791976 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110001.3 NP_001103471.1 1 - 2 151 110
GenBank mRNA BC120201.1 - 1 - 2 194 110

View Full Product Details