Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C14H8orf82
Entrez Gene ID
618766
Gene Name
chromosome 14 C8orf82 homolog
Gene Aliases
-
Chromosome Location
Chr.14: 416907 - 419447 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110184.1 NP_001103654.1 1 - 2 181 88
GenBank mRNA BC134700.1 - 1 - 2 181 88

View Full Product Details