Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXCL12
Entrez Gene ID
613811
Gene Name
C-X-C motif chemokine ligand 12
Gene Aliases
SDF1
Chromosome Location
Chr.28: 45022335 - 45051626 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113174.1 NP_001106645.1 1 - 2 95 71
GenBank mRNA BC119979.1 - 1 - 2 95 71
KP057086.1 - 1 - 2 59 71

View Full Product Details