Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CMC2
Entrez Gene ID
614078
Gene Name
C-X9-C motif containing 2
Gene Aliases
C18H16orf61
Chromosome Location
Chr.18: 7686031 - 7700190 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113175.2 NP_001106646.1 1 - 1 131 134
GenBank mRNA BC111685.1 - 1 - 1 132 134

View Full Product Details