Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHST2
Entrez Gene ID
615978
Gene Name
carbohydrate sulfotransferase 2
Gene Aliases
-
Chromosome Location
Chr.1: 125870582 - 125875385 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113769.1 NP_001107241.1 1 - 1 3153 125

View Full Product Details