Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOP10
Entrez Gene ID
616675
Gene Name
NOP10 ribonucleoprotein
Gene Aliases
NOLA3
Chromosome Location
Chr.10: 28299516 - 28303026 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114521.1 NP_001107993.1 1 - 2 104 57
GenBank mRNA BC103378.1 - 1 - 2 104 57

View Full Product Details