Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEIG1
Entrez Gene ID
617353
Gene Name
meiosis/spermiogenesis associated 1
Gene Aliases
-
Chromosome Location
Chr.13: 29595904 - 29612566 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114524.2 NP_001107996.2 1 - 2 185 70
GenBank mRNA BC109557.1 - 1 - 2 145 70
DT860200.1 - 1 - 2 177 70
EV609294.1 - 1 - 2 188 70

View Full Product Details