Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRNP27
Entrez Gene ID
790677
Gene Name
small nuclear ribonucleoprotein U4/U6.U5 subunit 27
Gene Aliases
RY1
Chromosome Location
Chr.11: 68242066 - 68249965 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098162.2 NP_001091631.1 5 - 6 439 86
GenBank mRNA BC133447.1 - 5 - 6 440 86

View Full Product Details