Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EGFL8
Entrez Gene ID
782820
Gene Name
EGF like domain multiple 8
Gene Aliases
NG3
Chromosome Location
Chr.23: 27221540 - 27224316 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099187.2 NP_001092657.1 3 - 4 303 125
GenBank mRNA BC146141.1 - 3 - 4 304 125

View Full Product Details