Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CT47B1
Entrez Gene ID
787180
Gene Name
cancer/testis antigen family 147, member B1
Gene Aliases
-
Chromosome Location
Chr.X: 5568549 - 5572125 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099202.1 NP_001092672.1 1 - 2 855 82
GenBank mRNA BC146048.1 - 1 - 2 855 82

View Full Product Details