Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNMD
Entrez Gene ID
781292
Gene Name
tenomodulin
Gene Aliases
-
Chromosome Location
Chr.X: 49576266 - 49597606 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099948.1 NP_001093418.1 6 - 7 748 86
GenBank mRNA EF608502.1 - 6 - 7 748 86

View Full Product Details