Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11H2orf73
Entrez Gene ID
782317
Gene Name
chromosome 11 C2orf73 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 37034101 - 37061615 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101283.2 NP_001094753.1 2 - 3 297 87
GenBank mRNA BC149887.1 - 2 - 3 298 87

View Full Product Details