Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMAN2
Entrez Gene ID
790870
Gene Name
lectin, mannose binding 2
Gene Aliases
-
Chromosome Location
Chr.7: 38829325 - 38849952 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101309.1 NP_001094779.1 3 - 4 452 75
GenBank mRNA BC149134.1 - 3 - 4 452 75

View Full Product Details