Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TNFRSF10D
- Entrez Gene ID
-
782274
- Gene Name
- tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Unknown: 91178 - 101849 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001102327.1 |
|
- |
1918 |
74 |
GenBank mRNA |
BC148939.1 |
- |
- |
1918 |
74 |
View Full Product Details