Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGALS3
Entrez Gene ID
786492
Gene Name
galectin 3
Gene Aliases
-
Chromosome Location
Chr.10: 67611548 - 67629447 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102341.2 NP_001095811.1 1 - 2 98 72
GenBank mRNA BC148136.1 - 1 - 2 99 72

View Full Product Details