Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD7
Entrez Gene ID
100124502
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
-
Chromosome Location
Chr.25: 28204658 - 28215127 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102545.1 NP_001096015.1 1 - 2 318 117
GenBank mRNA BC134499.1 - 1 - 2 318 117

View Full Product Details