Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD14
Entrez Gene ID
785513
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
-
Chromosome Location
Chr.19: 36526568 - 36544631 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105018.1 NP_001098488.1 4 - 5 1059 116
GenBank mRNA BC146255.1 - 4 - 5 1059 116

View Full Product Details