Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B4GALT2
Entrez Gene ID
100125390
Gene Name
beta-1,4-galactosyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.3: 102069525 - 102079822 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105030.2 NP_001098500.1 2 - 3 527 70
GenBank mRNA BC150042.1 - 2 - 3 528 70

View Full Product Details