Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIMAP7
Entrez Gene ID
100125415
Gene Name
GTPase, IMAP family member 7
Gene Aliases
-
Chromosome Location
Chr.4: 113276469 - 113287210 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105034.2 NP_001098504.1 1 - 2 47 115
GenBank mRNA BC149471.1 - 1 - 2 48 115

View Full Product Details